missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2,4,6-triiodoisophthaloyl Dichloride 98.0+%, TCI America™
$290.79 - $1010.32
Chemical Identifiers
| CAS | 37441-29-5 |
|---|---|
| Molecular Formula | C8H2Cl2I3NO2 |
| Molecular Weight (g/mol) | 595.722 |
| MDL Number | MFCD00270836 |
| InChI Key | FBJVWRITWDYUAC-UHFFFAOYSA-N |
| PubChem CID | 9894858 |
| IUPAC Name | 5-amino-2,4,6-triiodobenzene-1,3-dicarbonyl chloride |
| SMILES | C1(=C(C(=C(C(=C1I)N)I)C(=O)Cl)I)C(=O)Cl |
Chemical Identifiers
| 37441-29-5 | |
| 595.722 | |
| FBJVWRITWDYUAC-UHFFFAOYSA-N | |
| 5-amino-2,4,6-triiodobenzene-1,3-dicarbonyl chloride |
| C8H2Cl2I3NO2 | |
| MFCD00270836 | |
| 9894858 | |
| C1(=C(C(=C(C(=C1I)N)I)C(=O)Cl)I)C(=O)Cl |
Specifications
| 37441-29-5 | |
| C8H2Cl2I3NO2 | |
| 5 g | |
| FBJVWRITWDYUAC-UHFFFAOYSA-N | |
| 5-amino-2,4,6-triiodobenzene-1,3-dicarbonyl chloride | |
| 9894858 | |
| ≥98.0% (T) | |
| 5-Amino-2,4,6-triiodoisophthaloyl Dichloride |
| 231°C | |
| MFCD00270836 | |
| 3077 | |
| C1(=C(C(=C(C(=C1I)N)I)C(=O)Cl)I)C(=O)Cl | |
| 595.722 | |
| 595.72 | |
| Crystalline Powder |
Safety and Handling
TSCA : No