missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylphenol, 97%, Thermo Scientific Chemicals

$187.12 - $187.12

Chemical Identifiers

CAS 2835-95-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00043922
InChI Key DBFYESDCPWWCHN-UHFFFAOYSA-N
Synonym 5-amino-o-cresol, 4-amino-2-hydroxytoluene, 3-hydroxy-4-methylaniline, phenol, 5-amino-2-methyl, 3-amino-6-methylphenol, 6-methyl-3-aminophenol, unii-ubb8xeb10b, 5-amino-2-methyl-phenol, 5-amino-2-methyl phenol, ccris 4582
PubChem CID 17818
IUPAC Name 5-amino-2-methylphenol
SMILES CC1=C(C=C(C=C1)N)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6625222
View Documents
Thermo Scientific Chemicals
H6625222
100 g
Each for $187.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2835-95-2
123.155
DBFYESDCPWWCHN-UHFFFAOYSA-N
17818
CC1=C(C=C(C=C1)N)O
C7H9NO
MFCD00043922
5-amino-o-cresol, 4-amino-2-hydroxytoluene, 3-hydroxy-4-methylaniline, phenol, 5-amino-2-methyl, 3-amino-6-methylphenol, 6-methyl-3-aminophenol, unii-ubb8xeb10b, 5-amino-2-methyl-phenol, 5-amino-2-methyl phenol, ccris 4582
5-amino-2-methylphenol

Specifications

2835-95-2
Brown
MFCD00043922
2802317
DBFYESDCPWWCHN-UHFFFAOYSA-N
5-amino-2-methylphenol
17818
97%
5-Amino-2-methylphenol
160°C to 162°C
C7H9NO
100 g
5-amino-o-cresol, 4-amino-2-hydroxytoluene, 3-hydroxy-4-methylaniline, phenol, 5-amino-2-methyl, 3-amino-6-methylphenol, 6-methyl-3-aminophenol, unii-ubb8xeb10b, 5-amino-2-methyl-phenol, 5-amino-2-methyl phenol, ccris 4582
CC1=C(C=C(C=C1)N)O
123.155
123.16
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-618-6

RTECSNumber : SJ6090000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only