missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylpyridine 98.0+%, TCI America™

$39.14 - $115.57

Chemical Identifiers

CAS 3430-14-6
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00833389
InChI Key UENBBJXGCWILBM-UHFFFAOYSA-N
Synonym 5-amino-2-methylpyridine, 3-amino-6-methylpyridine, 3-amino-6-picoline, 5-amino-2-picoline, 6-methyl-pyridin-3-ylamine, 2-methyl-5-aminopyridine, 6-methylpyridin-3-ylamine, 3-pyridinamine, 6-methyl, 6-methyl-3-pyridinamine, 6-methyl-3-pyridylamine
PubChem CID 243313
IUPAC Name 6-methylpyridin-3-amine
SMILES CC1=CC=C(N)C=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A20531G
View Documents
TCI America
A20531G
1 g N/A
Only null left
 
A20535G
View Documents
TCI America
A20535G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3430-14-6
108.14
UENBBJXGCWILBM-UHFFFAOYSA-N
243313
CC1=CC=C(N)C=N1
C6H8N2
MFCD00833389
5-amino-2-methylpyridine, 3-amino-6-methylpyridine, 3-amino-6-picoline, 5-amino-2-picoline, 6-methyl-pyridin-3-ylamine, 2-methyl-5-aminopyridine, 6-methylpyridin-3-ylamine, 3-pyridinamine, 6-methyl, 6-methyl-3-pyridinamine, 6-methyl-3-pyridylamine
6-methylpyridin-3-amine

Specifications

3430-14-6
C6H8N2
1 g
5-amino-2-methylpyridine, 3-amino-6-methylpyridine, 3-amino-6-picoline, 5-amino-2-picoline, 6-methyl-pyridin-3-ylamine, 2-methyl-5-aminopyridine, 6-methylpyridin-3-ylamine, 3-pyridinamine, 6-methyl, 6-methyl-3-pyridinamine, 6-methyl-3-pyridylamine
CC1=CC=C(N)C=N1
108.14
108.14
Crystalline Powder
95°C
MFCD00833389
2811
UENBBJXGCWILBM-UHFFFAOYSA-N
6-methylpyridin-3-amine
243313
≥98.0% (GC,T)
5-Amino-2-methylpyridine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'