missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-naphthalenesulfonic Acid 97.0+%, TCI America™

$59.65 - $275.87

Chemical Identifiers

CAS 119-79-9
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00004030
InChI Key UWPJYQYRSWYIGZ-UHFFFAOYSA-N
Synonym 5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid
PubChem CID 8408
ChEBI CHEBI:44188
IUPAC Name 5-aminonaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A034625G
View Documents
TCI America
A034625G
25 g
Each for $59.65
Only null left
 
A0346500G
View Documents
TCI America
A0346500G
500 g
Each for $275.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

119-79-9
223.246
UWPJYQYRSWYIGZ-UHFFFAOYSA-N
8408
5-aminonaphthalene-2-sulfonic acid
C10H9NO3S
MFCD00004030
5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid
CHEBI:44188
C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N

Specifications

119-79-9
C10H9NO3S
25 g
UWPJYQYRSWYIGZ-UHFFFAOYSA-N
5-aminonaphthalene-2-sulfonic acid
8408
223.25
Crystalline Powder
Gray-Red
MFCD00004030
5-amino-2-naphthalenesulfonic acid, 1,6-cleve's acid, 1-aminonaphthalene-6-sulfonic acid, 2-naphthalenesulfonic acid, 5-amino, 1-naphthylamine-6-sulfonic acid, kyselina cleve, cleve's acid-1,6, 1-amino-6-sulfonaphthalene, 5-naphthylamine-2-sulfonic acid, cleve's beta-acid
C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C(=C1)N
223.246
CHEBI:44188
≥97.0% (T)
5-Amino-2-naphthalenesulfonic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0491

RTECSNumber : QK1285000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'