missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-naphthol 97.0+%, TCI America™

$1103.52 - $4147.52

Chemical Identifiers

CAS 86-97-5
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00035729
InChI Key FSBRKZMSECKELY-UHFFFAOYSA-N
PubChem CID 6865
IUPAC Name 5-aminonaphthalen-2-ol
SMILES C1=CC2=C(C=CC(=C2)O)C(=C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A03595G
View Documents
TCI America
A03595G
5 g
Each for $1,103.52
Only null left
 
A035925G
View Documents
TCI America
A035925G
25 g
Each for $4,147.52
Only null left
 
Description

Description

Specifications

Chemical Identifiers

86-97-5
159.188
FSBRKZMSECKELY-UHFFFAOYSA-N
5-aminonaphthalen-2-ol
C10H9NO
MFCD00035729
6865
C1=CC2=C(C=CC(=C2)O)C(=C1)N

Specifications

86-97-5
C10H9NO
5 g
C1=CC2=C(C=CC(=C2)O)C(=C1)N
159.188
159.19
Crystalline Powder
183°C
MFCD00035729
FSBRKZMSECKELY-UHFFFAOYSA-N
5-aminonaphthalen-2-ol
6865
≥97.0% (HPLC,T)
5-Amino-2-naphthol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0363

RTECSNumber : QL3330000

TSCA : Yes

SDS
Documents

Documents