missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-3-methyl-1-p-tolylpyrazole 97.0+%, TCI America™
Supplier: TCI America A095210G
Specifications
| 5-Amino-3-methyl-1-p-tolylpyrazole | |
| 118°C | |
| MFCD00020725 | |
| WQUNBEPKWJLYJD-UHFFFAOYSA-N | |
| 5-methyl-2-(4-methylphenyl)pyrazol-3-amine | |
| 112855 | |
| ≥97.0% (GC,T) |
| 62535-60-8 | |
| C11H13N3 | |
| 10 g | |
| CC1=CC=C(C=C1)N2C(=CC(=N2)C)N | |
| 187.246 | |
| 187.25 | |
| Crystalline Powder |
Chemical Identifiers
| 62535-60-8 | |
| 187.246 | |
| WQUNBEPKWJLYJD-UHFFFAOYSA-N | |
| 5-methyl-2-(4-methylphenyl)pyrazol-3-amine |
| C11H13N3 | |
| MFCD00020725 | |
| 112855 | |
| CC1=CC=C(C=C1)N2C(=CC(=N2)C)N |
Safety and Handling
TSCA : Yes