missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aminoindan, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 157350100

 View more versions of this product

Catalog No. AC157350100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Aminoindan
24425-40-9
100.0
>110°C
96% min. (GC)
C9H11N
10g
5-aminoindan, indan-5-amine, indan-5-ylamine, 5-indanamine, 5-aminohydrindene, 5-aminoindane, indane-5-ylamine, 1h-inden-5-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-5-ylamine, 6-aminoindane
LEWZOBYWGWKNCK-UHFFFAOYSA-N
2,3-dihydro-1H-inden-5-amine
90496
97%
97%
96.0
247.0°C to 249.0°C (745.0mmHg)
Authentic
Glass bottle
MFCD00003803
12, I, 511
Solubility in water: insoluble. Other solubilities: soluble in ether, acetone, benzene
C1CC2=C(C1)C=C(C=C2)N
133.19
133.19

Chemical Identifiers

24425-40-9
133.19
LEWZOBYWGWKNCK-UHFFFAOYSA-N
90496
C1CC2=C(C1)C=C(C=C2)N
C9H11N
MFCD00003803
5-aminoindan, indan-5-amine, indan-5-ylamine, 5-indanamine, 5-aminohydrindene, 5-aminoindane, indane-5-ylamine, 1h-inden-5-amine, 2,3-dihydro, 2,3-dihydro-1h-inden-5-ylamine, 6-aminoindane
2,3-dihydro-1H-inden-5-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Warning

EINECSNumber : 246-241-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only