missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1507418

 View more versions of this product

Catalog No. AAA1507418


Explore available promotions

Description

Description

5-Aminoisophthalic acid is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminoisophthalic acid is a useful biochemical for proteomics research.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

5-Aminoisophthalic acid
>300°C
MFCD00002522
2805628
Insoluble in water.
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
181.147
181.15
99-31-0
C8H7NO4
50 g
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
KBZFDRWPMZESDI-UHFFFAOYSA-N
5-aminobenzene-1,3-dicarboxylic acid
66833
95%

Chemical Identifiers

99-31-0
181.147
KBZFDRWPMZESDI-UHFFFAOYSA-N
66833
C1=C(C=C(C=C1C(=O)O)N)C(=O)O
C8H7NO4
MFCD00002522
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous
5-aminobenzene-1,3-dicarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 202-748-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only