Learn More
5-Aminoisophthalic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1507430
Description
5-Aminoisophthalic acid is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Aminoisophthalic acid is a useful biochemical for proteomics research.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
5-Aminoisophthalic acid | |
>300°C | |
MFCD00002522 | |
2805628 | |
Insoluble in water. | |
C1=C(C=C(C=C1C(=O)O)N)C(=O)O | |
181.147 | |
181.15 |
99-31-0 | |
C8H7NO4 | |
250 g | |
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous | |
KBZFDRWPMZESDI-UHFFFAOYSA-N | |
5-aminobenzene-1,3-dicarboxylic acid | |
66833 | |
95% |
Chemical Identifiers
99-31-0 | |
181.147 | |
KBZFDRWPMZESDI-UHFFFAOYSA-N | |
66833 | |
C1=C(C=C(C=C1C(=O)O)N)C(=O)O |
C8H7NO4 | |
MFCD00002522 | |
5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrous | |
5-aminobenzene-1,3-dicarboxylic acid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 202-748-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only