missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 103920050

 View more versions of this product

Catalog No. AC103920050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

156°C to 158°C
99%
98.5
White to Yellow
98.5% min. (HPLC)
Glass Bottle
MFCD00012869
5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
ZLHFONARZHCSET-UHFFFAOYSA-N
5-amino-4-oxopentanoic acid;hydrochloride
5 g
167.59
Crystalline Powder or Crystals
5-Aminolevulinic acid hydrochloride
5451-09-2
100.0
Authentic
C5H9NO3·HCl
H2NCH2COCH2CH2CO2H·HCl
15, 443
(5% in water) Clear colorless
C(CC(=O)O)C(=O)CN.Cl
167.59
123608
99%

Chemical Identifiers

5451-09-2
167.59
ZLHFONARZHCSET-UHFFFAOYSA-N
123608
C(CC(=O)O)C(=O)CN.Cl
C5H9NO3·HCl
MFCD00012869
5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
5-amino-4-oxopentanoic acid;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-679-5

RTECSNumber : OI1640000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only