missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Azacytosine 98.0+%, TCI America™
$109.98 - $439.92
Chemical Identifiers
| CAS | 931-86-2 |
|---|---|
| Molecular Formula | C3H4N4O |
| Molecular Weight (g/mol) | 112.09 |
| MDL Number | MFCD00006033,MFCD00149402,MFCD00051007 |
| InChI Key | MFEFTTYGMZOIKO-UHFFFAOYSA-N |
| Synonym | 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one |
| PubChem CID | 19956 |
| ChEBI | CHEBI:72474 |
| IUPAC Name | 6-amino-1,2-dihydro-1,3,5-triazin-2-one |
| SMILES | NC1=NC=NC(=O)N1 |
Chemical Identifiers
| 931-86-2 | |
| 112.09 | |
| MFEFTTYGMZOIKO-UHFFFAOYSA-N | |
| 19956 | |
| 6-amino-1,2-dihydro-1,3,5-triazin-2-one |
| C3H4N4O | |
| MFCD00006033,MFCD00149402,MFCD00051007 | |
| 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one | |
| CHEBI:72474 | |
| NC1=NC=NC(=O)N1 |
Specifications
| 931-86-2 | |
| C3H4N4O | |
| 5 g | |
| MFEFTTYGMZOIKO-UHFFFAOYSA-N | |
| 6-amino-1,2-dihydro-1,3,5-triazin-2-one | |
| 19956 | |
| 112.09 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00006033,MFCD00149402,MFCD00051007 | |
| 5-azacytosine, 1,3,5-triazin-2 1h-one, 4-amino, 4-amino-1,3,5-triazin-2 1h-one, 4-amino-1,3,5-triazin-2-ol, 4-amino-1,3,5-triazin-2-one, s-triazin-2 1h-one, 4-amino, s-triazin-2-ol, 4-amino, 2-amino-4-hydroxy-s-triazine, 6-amino-1,3,5-triazin-2 1h-one, 4-amino-s-triazin-2 1h-one | |
| NC1=NC=NC(=O)N1 | |
| 112.09 | |
| CHEBI:72474 | |
| ≥98.0% (T) | |
| 5-Azacytosine |
Safety and Handling
RTECSNumber : XZ2854300
TSCA : No