missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyl D-Glutamate 98.0+%, TCI America™

$54.06 - $76.43

Chemical Identifiers

CAS 2578-33-8
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00063111
InChI Key BGGHCRNCRWQABU-SNVBAGLBSA-N
Synonym h-d-glu obzl-oh, d-glutamic acid 5-benzyl ester, 5-benzyl d-glutamate, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, d-glutamicacid5-benzylester, d-glutamicacid5-benzylester, h-d-glu oblz-oh, h-d-glu oblz-oh, a-benzyl ester, a-benzyl ester
PubChem CID 6994624
IUPAC Name (2R)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39991G
View Documents
TCI America
B39991G
1 g
Each for $54.06
Only null left
 
B39995G
View Documents
TCI America
B39995G
5 g
Each for $76.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2578-33-8
237.255
BGGHCRNCRWQABU-SNVBAGLBSA-N
6994624
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
C12H15NO4
MFCD00063111
h-d-glu obzl-oh, d-glutamic acid 5-benzyl ester, 5-benzyl d-glutamate, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, d-glutamicacid5-benzylester, d-glutamicacid5-benzylester, h-d-glu oblz-oh, h-d-glu oblz-oh, a-benzyl ester, a-benzyl ester
(2R)-2-amino-5-oxo-5-phenylmethoxypentanoic acid

Specifications

2578-33-8
White
MFCD00063111
h-d-glu obzl-oh, d-glutamic acid 5-benzyl ester, 5-benzyl d-glutamate, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, 2r-2-amino-5-benzyloxy-5-oxopentanoic acid, d-glutamicacid5-benzylester, d-glutamicacid5-benzylester, h-d-glu oblz-oh, h-d-glu oblz-oh, a-benzyl ester, a-benzyl ester
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
237.255
237.26
Crystalline Powder
158°C
C12H15NO4
1 g
BGGHCRNCRWQABU-SNVBAGLBSA-N
(2R)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
6994624
≥98.0% (HPLC,T)
5-Benzyl D-Glutamate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'