missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Boc-amino)-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals
$343.32 - $1243.63
Chemical Identifiers
CAS | 503555-96-2 |
---|---|
Molecular Formula | C12H14ClNO4 |
Molecular Weight (g/mol) | 271.70 |
MDL Number | MFCD02682166 |
InChI Key | QNRXWUBPCIBQMX-UHFFFAOYSA-N |
Synonym | 5-tert-butoxycarbonyl amino-2-chlorobenzoic acid, n-boc-5-amino-2-chlorobenzoic acid, 5-boc-amino-2-chlorobenzoic acid, 5-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-5-2-methyl-2-propanyl oxy carbonyl amino benzoic acid, benzoic acid, 2-chloro-5-1,1-dimethylethoxy carbonyl amino-9ci |
PubChem CID | 22291441 |
IUPAC Name | 2-chloro-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid |
SMILES | CC(C)(C)OC(=O)NC1=CC(C(O)=O)=C(Cl)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH5230803
|
Thermo Scientific Chemicals
H5230803 |
1 g |
Each for $343.32
|
|
||||||
AAH5230806
|
Thermo Scientific Chemicals
H5230806 |
5 g |
Each for $1,243.63
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
503555-96-2 | |
271.70 | |
QNRXWUBPCIBQMX-UHFFFAOYSA-N | |
22291441 | |
CC(C)(C)OC(=O)NC1=CC(C(O)=O)=C(Cl)C=C1 |
C12H14ClNO4 | |
MFCD02682166 | |
5-tert-butoxycarbonyl amino-2-chlorobenzoic acid, n-boc-5-amino-2-chlorobenzoic acid, 5-boc-amino-2-chlorobenzoic acid, 5-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-5-2-methyl-2-propanyl oxy carbonyl amino benzoic acid, benzoic acid, 2-chloro-5-1,1-dimethylethoxy carbonyl amino-9ci | |
2-chloro-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid |
Specifications
503555-96-2 | |
MFCD02682166 | |
5-tert-butoxycarbonyl amino-2-chlorobenzoic acid, n-boc-5-amino-2-chlorobenzoic acid, 5-boc-amino-2-chlorobenzoic acid, 5-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-5-2-methyl-2-propanyl oxy carbonyl amino benzoic acid, benzoic acid, 2-chloro-5-1,1-dimethylethoxy carbonyl amino-9ci | |
CC(C)(C)OC(=O)NC1=CC(C(O)=O)=C(Cl)C=C1 | |
271.70 | |
271.7 | |
5-(Boc-amino)-2-chlorobenzoic acid |
C12H14ClNO4 | |
1 g | |
QNRXWUBPCIBQMX-UHFFFAOYSA-N | |
2-chloro-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid | |
22291441 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only