missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-(Boc-amino)valeric acid, 95%, Thermo Scientific™
$49.29 - $49.29
Chemical Identifiers
CAS | 27219-07-4 |
---|---|
Molecular Formula | C10H19NO4 |
Molecular Weight (g/mol) | 217.265 |
MDL Number | MFCD00076903 |
InChI Key | GFMRZAMDGJIWRB-UHFFFAOYSA-N |
Synonym | boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid |
PubChem CID | 545848 |
IUPAC Name | 5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
SMILES | CC(C)(C)OC(=O)NCCCCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6616803
|
Thermo Scientific Chemicals
H6616803 |
1 g |
Each for $49.29
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
27219-07-4 | |
217.265 | |
GFMRZAMDGJIWRB-UHFFFAOYSA-N | |
545848 | |
CC(C)(C)OC(=O)NCCCCC(=O)O |
C10H19NO4 | |
MFCD00076903 | |
boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid | |
5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Specifications
27219-07-4 | |
White | |
113°C (235°F) | |
(CH3)3COC(=O)NH(CH2)4CO2H | |
1 g | |
boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid | |
CC(C)(C)OC(=O)NCCCCC(=O)O | |
217.265 | |
217.27 | |
Powder |
48°C to 52°C | |
160°C to 168°C (0.8 mmHg) | |
C10H19NO4 | |
MFCD00076903 | |
2048862 | |
GFMRZAMDGJIWRB-UHFFFAOYSA-N | |
5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
545848 | |
95% | |
5-(Boc-amino)valeric acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures