missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1-methyl-1H-indole-3-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 400071-95-6 |
|---|---|
| Molecular Formula | C10H8BrNO2 |
| Molecular Weight (g/mol) | 254.08 |
| MDL Number | MFCD14706162 |
| InChI Key | WQZLFFJMRKTCMU-UHFFFAOYSA-N |
| Synonym | 5-bromo-1-methyl-1h-indole-3-carboxylic acid, 5-bromo-1-methyl-1h-indole-3-carboxylicacid, 1h-indole-3-carboxylic acid, 5-bromo-1-methyl, ksc230c9b, 5-bromo-1-methyl-1h-indole-3-carboxylic acid; |
| PubChem CID | 11032413 |
| IUPAC Name | 5-bromo-1-methylindole-3-carboxylic acid |
| SMILES | CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC431170010
|
Thermo Scientific Chemicals
431170010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC431170050
|
Thermo Scientific Chemicals
431170050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 400071-95-6 | |
| 254.08 | |
| WQZLFFJMRKTCMU-UHFFFAOYSA-N | |
| 11032413 | |
| CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O |
| C10H8BrNO2 | |
| MFCD14706162 | |
| 5-bromo-1-methyl-1h-indole-3-carboxylic acid, 5-bromo-1-methyl-1h-indole-3-carboxylicacid, 1h-indole-3-carboxylic acid, 5-bromo-1-methyl, ksc230c9b, 5-bromo-1-methyl-1h-indole-3-carboxylic acid; | |
| 5-bromo-1-methylindole-3-carboxylic acid |
Specifications
| 400071-95-6 | |
| 100.0 | |
| 96% min. (HPLC) | |
| C10H8BrNO2 | |
| 1g | |
| WQZLFFJMRKTCMU-UHFFFAOYSA-N | |
| 5-bromo-1-methylindole-3-carboxylic acid | |
| 11032413 | |
| 97% |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| MFCD14706162 | |
| 5-bromo-1-methyl-1h-indole-3-carboxylic acid, 5-bromo-1-methyl-1h-indole-3-carboxylicacid, 1h-indole-3-carboxylic acid, 5-bromo-1-methyl, ksc230c9b, 5-bromo-1-methyl-1h-indole-3-carboxylic acid; | |
| CN1C=C(C2=C1C=CC(=C2)Br)C(=O)O | |
| 254.08 | |
| 254.08 | |
| 5-Bromo-1-methyl-1H-indole-3-carboxylic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
RUO â Research Use Only