missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-1-pentene, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1121914

 View more versions of this product

Catalog No. AAA1121914


Explore available promotions

Description

Description

5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-1-pentene is widely utilized in the stereoselective synthesis of 7alpha-(3-carboxypropyl) estradiol. It plays an important role as starting material in the synthesis of a variety of compounds including DL-histrionicotoxin and benzophenone containing fatty acids.

Solubility
Immiscible with water.

Notes
Keep away from sources of ignition.
Specifications

Specifications

5-Bromo-1-pentene
1.259
30°C (86°F)
1.4639
MFCD0000-364
UN1993
5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
LPNANKDXVBMDKE-UHFFFAOYSA-N
5-bromopent-1-ene
70704
96%
1119-51-3
126°C to 127°C
C5H9Br
Br(CH2)3CH=CH2
25 g
506077
Immiscible with water.
C=CCCCBr
149.031
149.03

Chemical Identifiers

1119-51-3
149.031
LPNANKDXVBMDKE-UHFFFAOYSA-N
70704
C=CCCCBr
C5H9Br
MFCD0000-364
5-bromo-1-pentene, 1-pentene, 5-bromo, 1-bromo-4-pentene, 4-pentenyl bromide, 5-bromopentene, 5-bromo-pent-1-ene, 5-bromo-pentene, 5-bromopentene-1, 5-bromo-1-amylene, pent-4-enyl bromide
5-bromopent-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 214-281-4

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only