Learn More
5-Bromo-2,2-difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2366106
Description
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
Solubility
Miscible with hexane.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
5-Bromo-2,2-difluoro-1,3-benzodioxole | |
1.74 | |
C7H3BrF2O2 | |
MFCD00236212 | |
1425209 | |
Miscible with hexane. | |
C1=CC2=C(C=C1Br)OC(O2)(F)F | |
237 | |
237 |
33070-32-5 | |
78°C to 79°C (20 mmHg) | |
1.4967 | |
5 g | |
5-bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2h-1,3-benzodioxole, 5-bromo-2,2-difluorobenzo d 1,3 dioxole, 5-bromo-2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo-2,2-difluoro, 4-bromo-1,2-difluoromethylenedioxy benzene, 5-bromo-2,2-difluorobenzo d 1,3-dioxolene, acmc-209hy8, 5-bromo2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-5-bromo-1,3-benzodioxole | |
SZRHWHHXVXSGMT-UHFFFAOYSA-N | |
5-bromo-2,2-difluoro-1,3-benzodioxole | |
2736271 | |
97% |
Chemical Identifiers
33070-32-5 | |
237 | |
SZRHWHHXVXSGMT-UHFFFAOYSA-N | |
2736271 | |
C1=CC2=C(C=C1Br)OC(O2)(F)F |
C7H3BrF2O2 | |
MFCD00236212 | |
5-bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2h-1,3-benzodioxole, 5-bromo-2,2-difluorobenzo d 1,3 dioxole, 5-bromo-2,2-difluoro-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo-2,2-difluoro, 4-bromo-1,2-difluoromethylenedioxy benzene, 5-bromo-2,2-difluorobenzo d 1,3-dioxolene, acmc-209hy8, 5-bromo2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-5-bromo-1,3-benzodioxole | |
5-bromo-2,2-difluoro-1,3-benzodioxole |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only