missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2-chloroanisole, 97%, Thermo Scientific™
$57.36 - $57.36
Chemical Identifiers
CAS | 16817-43-9 |
---|---|
Molecular Formula | C7H6BrClO |
Molecular Weight (g/mol) | 221.48 |
MDL Number | MFCD00078659 |
InChI Key | UAMVKOTWSHJOSY-UHFFFAOYSA-N |
Synonym | 5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq |
PubChem CID | 2769567 |
IUPAC Name | 4-bromo-1-chloro-2-methoxybenzene |
SMILES | COC1=C(Cl)C=CC(Br)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5553303
|
Thermo Scientific Chemicals
H5553303 |
1 g |
Each for $57.36
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications5-Bromo-2-chloroanisole is used to produce 4-chloro-3-methoxy-benzonitrile at the temperature of 110°C. It will need solvent dimethylformamide
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
16817-43-9 | |
221.48 | |
UAMVKOTWSHJOSY-UHFFFAOYSA-N | |
2769567 | |
COC1=C(Cl)C=CC(Br)=C1 |
C7H6BrClO | |
MFCD00078659 | |
5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq | |
4-bromo-1-chloro-2-methoxybenzene |
Specifications
16817-43-9 | |
1.631 | |
113°C (235°F) | |
1.587 | |
1 g | |
Soluble in water. | |
COC1=C(Cl)C=CC(Br)=C1 | |
221.48 | |
221.48 | |
5-Bromo-2-chloroanisole |
27°C to 28°C | |
240°C to 245°C | |
C7H6BrClO | |
MFCD00078659 | |
5-bromo-2-chloroanisole, 5-bromo-2-chloroanisol, 4-chloro-3-methoxybromobenzene, 2-chloro-5-bromoanisole, 5-bromo-2-chloro-anisole, benzene, 4-bromo-1-chloro-2-methoxy, 4-bromo-1-chloro-2-methoxy-benzene, zlchem 582, pubchem2632, acmc-209dxq | |
UAMVKOTWSHJOSY-UHFFFAOYSA-N | |
4-bromo-1-chloro-2-methoxybenzene | |
2769567 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures