missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-chlorobenzoic Acid 98.0+%, TCI America™
$61.51 - $145.40
Chemical Identifiers
| CAS | 21739-92-4 |
|---|---|
| Molecular Formula | C7H4BrClO2 |
| Molecular Weight (g/mol) | 235.461 |
| MDL Number | MFCD00002415 |
| InChI Key | FGERXQWKKIVFQG-UHFFFAOYSA-N |
| Synonym | 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r |
| PubChem CID | 33127 |
| IUPAC Name | 5-bromo-2-chlorobenzoic acid |
| SMILES | C1=CC(=C(C=C1Br)C(=O)O)Cl |
Chemical Identifiers
| 21739-92-4 | |
| 235.461 | |
| FGERXQWKKIVFQG-UHFFFAOYSA-N | |
| 33127 | |
| C1=CC(=C(C=C1Br)C(=O)O)Cl |
| C7H4BrClO2 | |
| MFCD00002415 | |
| 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r | |
| 5-bromo-2-chlorobenzoic acid |
Specifications
| 21739-92-4 | |
| White-Yellow | |
| MFCD00002415 | |
| 2-chloro-5-bromobenzoic acid, benzoic acid, 5-bromo-2-chloro, 5-bromo-2-chlorobenzoicacid, 5-bromo-2-chloro benzoic acid, 5-bromo-2-chloro-benzoic acid, bromo 5--2-chlorobenzoic acid, pubchem3587, acmc-209fnm, 5-bromo-2-chorobenzoic acid, ksc204k3r | |
| C1=CC(=C(C=C1Br)C(=O)O)Cl | |
| 235.461 | |
| 235.46 | |
| Crystalline Powder |
| 158°C | |
| C7H4BrClO2 | |
| 10 g | |
| FGERXQWKKIVFQG-UHFFFAOYSA-N | |
| 5-bromo-2-chlorobenzoic acid | |
| 33127 | |
| ≥98.0% (GC,T) | |
| 5-Bromo-2-chlorobenzoic Acid |
Safety and Handling
TSCA : No