Learn More
5-Bromo-2-chloropyrimidine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1836006
Description
It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.
Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
5-Bromo-2-chloropyrimidine | |
77°C to 80°C | |
C4H2BrClN2 | |
5 g | |
2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine | |
XPGIBDJXEVAVTO-UHFFFAOYSA-N | |
5-bromo-2-chloropyrimidine | |
606665 | |
≥98% |
32779-36-5 | |
95°C (15 mmHg) | |
MFCD00483232 | |
507955 | |
Insoluble in water. Slightly soluble in DMSO and methanol. | |
ClC1=NC=C(Br)C=N1 | |
193.43 | |
193.43 |
Chemical Identifiers
32779-36-5 | |
193.43 | |
XPGIBDJXEVAVTO-UHFFFAOYSA-N | |
606665 | |
ClC1=NC=C(Br)C=N1 |
C4H2BrClN2 | |
MFCD00483232 | |
2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine | |
5-bromo-2-chloropyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only