missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Bromo-2'-deoxycytidine, MP Biomedicals™
5-Bromo-2'-deoxycytidine can be incorporated into DNA in the presence of DNA polymerase, replacing dCTP
$126.65 - $126.65
Chemical Identifiers
CAS | 1022-79-3 |
---|---|
Molecular Formula | C9H12BrN3O4 |
Molecular Weight (g/mol) | 306.116 |
InChI Key | KISUPFXQEHWGAR-RRKCRQDMSA-N |
Synonym | 5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy |
PubChem CID | 472175 |
IUPAC Name | 4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
SMILES | C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10016683
|
MP Biomedicals Inc
0210016683 |
250 mg |
N/A
|
|
|||||
Chemical Identifiers
1022-79-3 | |
306.116 | |
5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy | |
4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
C9H12BrN3O4 | |
KISUPFXQEHWGAR-RRKCRQDMSA-N | |
472175 | |
C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O |
Specifications
1022-79-3 | |
C9H12BrN3O4 | |
5-bromo-2'-deoxycytidine, 5-bromodeoxycytidine, 4-amino-5-bromo-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, unii-6m3tp1aibh, brcdr, bromodeoxycytidine, 6m3tp1aibh, bcdr, 5-bromo-2'-deoxycytidine monohydrate, cytidine, 5-bromo-2'-deoxy | |
C1C(C(OC1N2C=C(C(=NC2=O)N)Br)CO)O | |
306.116 | |
306.1 (Anhydrous) | |
5-Bromo-2'-deoxycytidine monohydrate |
White | |
250 mg | |
KISUPFXQEHWGAR-RRKCRQDMSA-N | |
4-amino-5-bromo-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one | |
472175 | |
Powder |
Safety and Handling
EINECSNumber : 213-824-2
Recommended Storage : Store at 0°C. Store Desiccated.