Learn More
5-Bromo-2-(difluoromethyl)pyridine, 95%, Thermo Scientific Chemicals
$363.66 - $363.66
Chemical Identifiers
| CAS | 845827-13-6 |
|---|---|
| Molecular Formula | C6H4BrF2N |
| Molecular Weight (g/mol) | 208.01 |
| MDL Number | MFCD11977429 |
| InChI Key | QXLZRIGSWWQOLG-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-difluoromethyl pyridine, pyridine, 5-bromo-2-difluoromethyl, 2-difluoromethyl-5-bromopyridine, pyridine,5-bromo-2-difluoromethyl, 3-bromo-6-difluoromethyl pyridine, 2-bis fluoranyl methyl-5-bromanyl-pyridine, 5-bromo-alpha,alpha-difluoro-2-picoline |
| PubChem CID | 53415062 |
| IUPAC Name | 5-bromo-2-(difluoromethyl)pyridine |
| SMILES | FC(F)C1=NC=C(Br)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6629703
|
Thermo Scientific Chemicals
H6629703 |
1 g |
Each for $363.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 845827-13-6 | |
| 208.01 | |
| QXLZRIGSWWQOLG-UHFFFAOYSA-N | |
| 53415062 | |
| FC(F)C1=NC=C(Br)C=C1 |
| C6H4BrF2N | |
| MFCD11977429 | |
| 5-bromo-2-difluoromethyl pyridine, pyridine, 5-bromo-2-difluoromethyl, 2-difluoromethyl-5-bromopyridine, pyridine,5-bromo-2-difluoromethyl, 3-bromo-6-difluoromethyl pyridine, 2-bis fluoranyl methyl-5-bromanyl-pyridine, 5-bromo-alpha,alpha-difluoro-2-picoline | |
| 5-bromo-2-(difluoromethyl)pyridine |
Specifications
| 845827-13-6 | |
| White to Yellow | |
| C6H4BrF2N | |
| 1 g | |
| 5-bromo-2-difluoromethyl pyridine, pyridine, 5-bromo-2-difluoromethyl, 2-difluoromethyl-5-bromopyridine, pyridine,5-bromo-2-difluoromethyl, 3-bromo-6-difluoromethyl pyridine, 2-bis fluoranyl methyl-5-bromanyl-pyridine, 5-bromo-alpha,alpha-difluoro-2-picoline | |
| FC(F)C1=NC=C(Br)C=C1 | |
| 208.01 | |
| 208 | |
| Solid |
| 35°C to 40°C | |
| 89°C (192°F) | |
| MFCD11977429 | |
| UN2811 | |
| QXLZRIGSWWQOLG-UHFFFAOYSA-N | |
| 5-bromo-2-(difluoromethyl)pyridine | |
| 53415062 | |
| 95% | |
| 5-Bromo-2-(difluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only