Learn More
5-Bromo-2-hydroxy-4-methylpyridine, 97%
CAS: 164513-38-6 | C6H6BrNO | 188.024 g/mol
$100.45 - $370.71
Chemical Identifiers
| CAS | 164513-38-6 |
|---|---|
| Molecular Formula | C6H6BrNO |
| Molecular Weight (g/mol) | 188.024 |
| MDL Number | MFCD03427664 |
| InChI Key | BYGNXKDNIFHZHQ-UHFFFAOYSA-N |
| Synonym | 5-bromo-2-hydroxy-4-methylpyridine, 5-bromo-4-methylpyridin-2-ol, 5-bromo-2-hydroxy-4-picoline, 5-bromo-4-methyl-2 1h-pyridinone, 5-bromo-4-methyl-2-pyridinol, 2-hydroxy-4-methyl-5-bromopyridine, 5-bromo-4-methyl-pyridin-2-ol, pubchem6564, acmc-209dre, ksc496a8d |
| PubChem CID | 819937 |
| IUPAC Name | 5-bromo-4-methyl-1H-pyridin-2-one |
| SMILES | CC1=CC(=O)NC=C1Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6467406
|
Thermo Scientific Chemicals
H6467406 |
5 g |
Each for $100.45
|
|
|||||
|
AAH6467414
|
Thermo Scientific Chemicals
H6467414 |
25 g |
Each for $370.71
|
|
|||||
Description
It is used for medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used for medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
| 164513-38-6 | |
| 188.024 | |
| BYGNXKDNIFHZHQ-UHFFFAOYSA-N | |
| 819937 | |
| CC1=CC(=O)NC=C1Br |
| C6H6BrNO | |
| MFCD03427664 | |
| 5-bromo-2-hydroxy-4-methylpyridine, 5-bromo-4-methylpyridin-2-ol, 5-bromo-2-hydroxy-4-picoline, 5-bromo-4-methyl-2 1h-pyridinone, 5-bromo-4-methyl-2-pyridinol, 2-hydroxy-4-methyl-5-bromopyridine, 5-bromo-4-methyl-pyridin-2-ol, pubchem6564, acmc-209dre, ksc496a8d | |
| 5-bromo-4-methyl-1H-pyridin-2-one |
Specifications
| 164513-38-6 | |
| White | |
| MFCD03427664 | |
| 6798531 | |
| Slightly soluble in water. | |
| CC1=CC(=O)NC=C1Br | |
| 188.024 | |
| 188.02 | |
| Solid |
| 198°C to 202°C | |
| C6H6BrNO | |
| 5 g | |
| 5-bromo-2-hydroxy-4-methylpyridine, 5-bromo-4-methylpyridin-2-ol, 5-bromo-2-hydroxy-4-picoline, 5-bromo-4-methyl-2 1h-pyridinone, 5-bromo-4-methyl-2-pyridinol, 2-hydroxy-4-methyl-5-bromopyridine, 5-bromo-4-methyl-pyridin-2-ol, pubchem6564, acmc-209dre, ksc496a8d | |
| BYGNXKDNIFHZHQ-UHFFFAOYSA-N | |
| 5-bromo-4-methyl-1H-pyridin-2-one | |
| 819937 | |
| 97% | |
| 5-Bromo-2-hydroxy-4-methylpyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only