missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-2-hydroxybenzyl alcohol, 98%, Thermo Scientific Chemicals

$148.28 - $148.28

Chemical Identifiers

CAS 2316-64-5
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.04
MDL Number MFCD00004618
InChI Key KNKRHSVKIORZQB-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
PubChem CID 75342
IUPAC Name 4-bromo-2-(hydroxymethyl)phenol
SMILES OCC1=CC(Br)=CC=C1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC331480050
SDS View Documents
Thermo Scientific Chemicals
331480050
5 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2316-64-5
203.04
KNKRHSVKIORZQB-UHFFFAOYSA-N
75342
OCC1=CC(Br)=CC=C1O
C7H7BrO2
MFCD00004618
5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
4-bromo-2-(hydroxymethyl)phenol

Specifications

2316-64-5
100.0
White to Brown
97.5% min. (GC)
C7H7BrO2
MFCD00004618
06, 893
5-bromo-2-hydroxybenzyl alcohol, 4-bromo-2-hydroxymethyl phenol, bromosaligenin, 5-bromosalicyl alcohol, bromsalizol, 5-bromosaligenin, 5-bromo-2-hydroxybenzylalcohol, 2-hydroxymethyl-4-bromophenol, benzenemethanol, 5-bromo-2-hydroxy, unii-y1uz53815e
KNKRHSVKIORZQB-UHFFFAOYSA-N
4-bromo-2-(hydroxymethyl)phenol
75342
98%
5-Bromo-2-hydroxybenzyl alcohol, 98%
97.5
106.0°C to 111.0°C
Authentic
Glass bottle
BrC6H3(OH)CH2OH
5 g
12, 1460
Solubility in water: insoluble in water. Other solubilities: soluble in acetone,benzene,chloroform
OCC1=CC(Br)=CC=C1O
203.04
203.04
Crystalline Powder or Flakes
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-026-0

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only