missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-methoxypyridine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 13472-85-0 |
|---|---|
| Molecular Formula | C6H6BrNO |
| Molecular Weight (g/mol) | 188.02 |
| MDL Number | MFCD01318952 |
| InChI Key | XADICJHFELMBGX-UHFFFAOYSA-N |
| Synonym | 2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408 |
| PubChem CID | 2734895 |
| IUPAC Name | 5-bromo-2-methoxypyridine |
| SMILES | COC1=NC=C(C=C1)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC376880250
|
Thermo Scientific Chemicals
376880250 |
25mL | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 13472-85-0 | |
| 188.02 | |
| XADICJHFELMBGX-UHFFFAOYSA-N | |
| 2734895 | |
| COC1=NC=C(C=C1)Br |
| C6H6BrNO | |
| MFCD01318952 | |
| 2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408 | |
| 5-bromo-2-methoxypyridine |
Specifications
| 13472-85-0 | |
| 100.0 | |
| 80.0°C (12.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5540 to 1.5560 | |
| 25mL | |
| 2-methoxy-5-bromopyridine, pyridine, 5-bromo-2-methoxy, 3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-pyridine, 2-methyloxy-5-bromopyridine, 5-bromo-2-methoxy pyridine, 2-methoxy-5-bromo-pyridine, 2-methyloxy-5-bromo pyridine, zlchem 355, pubchem2408 | |
| COC1=NC=C(C=C1)Br | |
| 188.02 | |
| 188.02 | |
| 5-Bromo-2-methoxypyridine |
| 96.0 | |
| 1.4530g/mL | |
| 96°C | |
| 96% min. (GC) | |
| C6H6BrNO | |
| MFCD01318952 | |
| 1.453 | |
| XADICJHFELMBGX-UHFFFAOYSA-N | |
| 5-bromo-2-methoxypyridine | |
| 2734895 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only