Learn More
5-Bromo-2-methylphenol, 95%
CAS: 36138-76-8 | C7H7BrO | 187.036 g/mol
$65.67 - $368.95
Chemical Identifiers
| CAS | 36138-76-8 |
|---|---|
| Molecular Formula | C7H7BrO |
| Molecular Weight (g/mol) | 187.036 |
| MDL Number | MFCD08061906 |
| InChI Key | OONJCAWRVJDVBB-UHFFFAOYSA-N |
| Synonym | 5-bromo-o-cresol, phenol, 5-bromo-2-methyl, 4-bromo-2-hydroxytoluene, 5-bromo-2-methyl phenol, 5-bromo-2-methyl-phenol, 3-bromo-6-methylphenol, 2-hydroxy-4-bromotoluene, 5-brom-2-kresol, zlchem 468, pubchem4132 |
| PubChem CID | 12648404 |
| IUPAC Name | 5-bromo-2-methylphenol |
| SMILES | CC1=C(C=C(C=C1)Br)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2695603
|
Thermo Scientific Chemicals
H2695603 |
1 g |
N/A
|
|
|||||
|
AAH2695609
|
Thermo Scientific Chemicals
H2695609 |
10 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 36138-76-8 | |
| 187.036 | |
| OONJCAWRVJDVBB-UHFFFAOYSA-N | |
| 12648404 | |
| CC1=C(C=C(C=C1)Br)O |
| C7H7BrO | |
| MFCD08061906 | |
| 5-bromo-o-cresol, phenol, 5-bromo-2-methyl, 4-bromo-2-hydroxytoluene, 5-bromo-2-methyl phenol, 5-bromo-2-methyl-phenol, 3-bromo-6-methylphenol, 2-hydroxy-4-bromotoluene, 5-brom-2-kresol, zlchem 468, pubchem4132 | |
| 5-bromo-2-methylphenol |
Specifications
| 36138-76-8 | |
| C7H7BrO | |
| 1 g | |
| OONJCAWRVJDVBB-UHFFFAOYSA-N | |
| 5-bromo-2-methylphenol | |
| 12648404 | |
| 95% |
| 77°C to 80°C | |
| MFCD08061906 | |
| 5-bromo-o-cresol, phenol, 5-bromo-2-methyl, 4-bromo-2-hydroxytoluene, 5-bromo-2-methyl phenol, 5-bromo-2-methyl-phenol, 3-bromo-6-methylphenol, 2-hydroxy-4-bromotoluene, 5-brom-2-kresol, zlchem 468, pubchem4132 | |
| CC1=C(C=C(C=C1)Br)O | |
| 187.036 | |
| 187.04 | |
| 5-Bromo-2-methylphenol |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only