Learn More
5-Bromo-2-nitrophenol, 96%, Thermo Scientific Chemicals
$124.75 - $478.92
Chemical Identifiers
CAS | 27684-84-0 |
---|---|
Molecular Formula | C6H4BrNO3 |
Molecular Weight (g/mol) | 218.006 |
MDL Number | MFCD03095027 |
InChI Key | DTWHNSNSUBKGTC-UHFFFAOYSA-N |
Synonym | 5-bromo-2-nitrophenol, phenol, 5-bromo-2-nitro, 3-bromo-6-nitrophenol, 4-bromo-2-hydroxynitrobenzene, 2-nitro-5-bromophenol, 5-bromo-2-nitro-phenol, phenol,5-bromo-2-nitro, pubchem16073, acmc-1cct0, 5-bromanyl-2-nitro-phenol |
PubChem CID | 13970496 |
SMILES | C1=CC(=C(C=C1Br)O)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6496606
|
Thermo Scientific Chemicals
H6496606 |
5 g |
Each for $124.75
|
|
|||||
AAH6496614
|
Thermo Scientific Chemicals
H6496614 |
25 g |
Each for $478.92
|
|
|||||
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate.
Solubility
Soluble in ethyl acetate and methanol.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
27684-84-0 | |
218.006 | |
DTWHNSNSUBKGTC-UHFFFAOYSA-N | |
13970496 |
C6H4BrNO3 | |
MFCD03095027 | |
5-bromo-2-nitrophenol, phenol, 5-bromo-2-nitro, 3-bromo-6-nitrophenol, 4-bromo-2-hydroxynitrobenzene, 2-nitro-5-bromophenol, 5-bromo-2-nitro-phenol, phenol,5-bromo-2-nitro, pubchem16073, acmc-1cct0, 5-bromanyl-2-nitro-phenol | |
C1=CC(=C(C=C1Br)O)[N+](=O)[O-] |
Specifications
27684-84-0 | |
Yellow | |
MFCD03095027 | |
2329085 | |
Soluble in ethyl acetate and methanol. | |
C1=CC(=C(C=C1Br)O)[N+](=O)[O-] | |
13970496 | |
96% | |
5-Bromo-2-nitrophenol |
40°C to 41°C | |
C6H4BrNO3 | |
5 g | |
5-bromo-2-nitrophenol, phenol, 5-bromo-2-nitro, 3-bromo-6-nitrophenol, 4-bromo-2-hydroxynitrobenzene, 2-nitro-5-bromophenol, 5-bromo-2-nitro-phenol, phenol,5-bromo-2-nitro, pubchem16073, acmc-1cct0, 5-bromanyl-2-nitro-phenol | |
DTWHNSNSUBKGTC-UHFFFAOYSA-N | |
218.006 | |
218.01 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only