missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromo-4-methyl-2-phenyl-1,3-oxazole, 95%, Thermo Scientific™

Chemical Identifiers

CAS 21354-98-3
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.08
MDL Number MFCD11109319
InChI Key QHQQHNFHCQSTBJ-UHFFFAOYSA-N
Synonym 5-bromo-4-methyl-2-phenyloxazole, 4-methyl-2-phenyl-5-bromooxazole, oxazole,5-bromo-4-methyl-2-phenyl, 5-bromo-4-methyl-1,3-oxazol-2-yl benzene
PubChem CID 15328586
IUPAC Name 5-bromo-4-methyl-2-phenyl-1,3-oxazole
SMILES CC1=C(Br)OC(=N1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC18110DA
SDS View Documents
Thermo Scientific Chemicals
CC18110DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC18110DE
SDS View Documents
Thermo Scientific Chemicals
CC18110DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

21354-98-3
238.08
QHQQHNFHCQSTBJ-UHFFFAOYSA-N
15328586
CC1=C(Br)OC(=N1)C1=CC=CC=C1
C10H8BrNO
MFCD11109319
5-bromo-4-methyl-2-phenyloxazole, 4-methyl-2-phenyl-5-bromooxazole, oxazole,5-bromo-4-methyl-2-phenyl, 5-bromo-4-methyl-1,3-oxazol-2-yl benzene
5-bromo-4-methyl-2-phenyl-1,3-oxazole

Specifications

21354-98-3
100.0
Amber Glass Bottle
MFCD11109319
5-bromo-4-methyl-2-phenyloxazole, 4-methyl-2-phenyl-5-bromooxazole, oxazole,5-bromo-4-methyl-2-phenyl, 5-bromo-4-methyl-1,3-oxazol-2-yl benzene
CC1=C(Br)OC(=N1)C1=CC=CC=C1
238.08
238.08
5-Bromo-4-methyl-2-phenyl-1, 3-oxazole
94.5
95%
C10H8BrNO
1g
QHQQHNFHCQSTBJ-UHFFFAOYSA-N
5-bromo-4-methyl-2-phenyl-1,3-oxazole
15328586
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'