missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromobenzo[b]furan-2-carbaldehyde, 97%, Thermo Scientific™

Chemical Identifiers

CAS 23145-16-6
Molecular Formula C9H5BrO2
Molecular Weight (g/mol) 225.041
InChI Key UPEGFMDITWHVHV-UHFFFAOYSA-N
Synonym 5-bromobenzofuran-2-carbaldehyde, 5-bromobenzo b furan-2-carbaldehyde, 5-bromo-2-formylbenzofuran, 5-bromo-2-benzofurancarboxaldehyde, 5-bromo-1-benzofuran-2-carboxaldehyde, 2-benzofurancarboxaldehyde,5-bromo, 5-bromobenzo b furan-2-carboxaldehyde, pubchem10165, acmc-1cjl8, 5-bromo-benzofuran-2-carbaldehyde
PubChem CID 2795535
IUPAC Name 5-bromo-1-benzofuran-2-carbaldehyde
SMILES C1=CC2=C(C=C1Br)C=C(O2)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC31204CB
SDS View Documents
Thermo Scientific Chemicals
CC31204CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31204DA
SDS View Documents
Thermo Scientific Chemicals
CC31204DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31204DE
SDS View Documents
Thermo Scientific Chemicals
CC31204DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

23145-16-6
225.041
5-bromobenzofuran-2-carbaldehyde, 5-bromobenzo b furan-2-carbaldehyde, 5-bromo-2-formylbenzofuran, 5-bromo-2-benzofurancarboxaldehyde, 5-bromo-1-benzofuran-2-carboxaldehyde, 2-benzofurancarboxaldehyde,5-bromo, 5-bromobenzo b furan-2-carboxaldehyde, pubchem10165, acmc-1cjl8, 5-bromo-benzofuran-2-carbaldehyde
5-bromo-1-benzofuran-2-carbaldehyde
C9H5BrO2
UPEGFMDITWHVHV-UHFFFAOYSA-N
2795535
C1=CC2=C(C=C1Br)C=C(O2)C=O

Specifications

23145-16-6
Amber Glass Bottle
250mg
UPEGFMDITWHVHV-UHFFFAOYSA-N
5-bromo-1-benzofuran-2-carbaldehyde
2795535
97%
97%
C9H5BrO2
5-bromobenzofuran-2-carbaldehyde, 5-bromobenzo b furan-2-carbaldehyde, 5-bromo-2-formylbenzofuran, 5-bromo-2-benzofurancarboxaldehyde, 5-bromo-1-benzofuran-2-carboxaldehyde, 2-benzofurancarboxaldehyde,5-bromo, 5-bromobenzo b furan-2-carboxaldehyde, pubchem10165, acmc-1cjl8, 5-bromo-benzofuran-2-carbaldehyde
C1=CC2=C(C=C1Br)C=C(O2)C=O
225.041
225.04
5-Bromo-1-benzofuran-2-carbaldehyde
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'