missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromobenzo[b]furan-2-carbonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 62878-96-0
Molecular Formula C9H4BrClO2
Molecular Weight (g/mol) 259.483
InChI Key FGPAEHRRVKLUHA-UHFFFAOYSA-N
Synonym 5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride
PubChem CID 2795534
IUPAC Name 5-bromo-1-benzofuran-2-carbonyl chloride
SMILES C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC31202CB
SDS View Documents
Thermo Scientific Chemicals
CC31202CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31202DA
SDS View Documents
Thermo Scientific Chemicals
CC31202DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31202DE
SDS View Documents
Thermo Scientific Chemicals
CC31202DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

62878-96-0
259.483
5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride
5-bromo-1-benzofuran-2-carbonyl chloride
C9H4BrClO2
FGPAEHRRVKLUHA-UHFFFAOYSA-N
2795534
C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl

Specifications

62878-96-0
Amber Glass Bottle
250mg
FGPAEHRRVKLUHA-UHFFFAOYSA-N
5-bromo-1-benzofuran-2-carbonyl chloride
2795534
95%
95%
C9H4BrClO2
5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride
C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl
259.483
259.49
5-Bromobenzo[b]furan-2-carbonyl chloride
SDS
missing translation for 'documents'

missing translation for 'documents'