missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
5-Bromobenzo[b]furan-2-carboxylic acid 97+%, Thermo Scientific™
Chemical Identifiers
CAS |
10242-11-2 |
Molecular Formula |
C9H5BrO3 |
Molecular Weight (g/mol) |
241.04 |
InChI Key |
QKUWZCOVKRUXKX-UHFFFAOYSA-N |
Synonym |
5-bromobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 5-bromo, 5-bromobenzo b furan-2-carboxylic acid, 5-bromo-2-carboxy-1-benzofuran, 5-bromo-2-benzofurancarboxylic acid, 2-benzofurancarboxylicacid, 5-bromo, 5-bromo-benzofuran-2-carboxylic acid, acmc-1bxoo, buttpark 34\07-74, 5-bromo benzo furan 2-carboxylic acid |
PubChem CID |
735247 |
IUPAC Name |
5-bromo-1-benzofuran-2-carboxylic acid |
SMILES |
C1=CC2=C(C=C1Br)C=C(O2)C(=O)O |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
10242-11-2 |
|
241.04 |
|
5-bromobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 5-bromo, 5-bromobenzo b furan-2-carboxylic acid, 5-bromo-2-carboxy-1-benzofuran, 5-bromo-2-benzofurancarboxylic acid, 2-benzofurancarboxylicacid, 5-bromo, 5-bromo-benzofuran-2-carboxylic acid, acmc-1bxoo, buttpark 34\07-74, 5-bromo benzo furan 2-carboxylic acid |
|
5-bromo-1-benzofuran-2-carboxylic acid |
|
C9H5BrO3 |
|
QKUWZCOVKRUXKX-UHFFFAOYSA-N |
|
735247 |
|
C1=CC2=C(C=C1Br)C=C(O2)C(=O)O |
Specifications
|
10242-11-2 |
|
Amber Glass Bottle |
|
1g |
|
QKUWZCOVKRUXKX-UHFFFAOYSA-N |
|
5-bromo-1-benzofuran-2-carboxylic acid |
|
735247 |
|
97% |
|
97% |
|
C9H5BrO3 |
|
5-bromobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 5-bromo, 5-bromobenzo b furan-2-carboxylic acid, 5-bromo-2-carboxy-1-benzofuran, 5-bromo-2-benzofurancarboxylic acid, 2-benzofurancarboxylicacid, 5-bromo, 5-bromo-benzofuran-2-carboxylic acid, acmc-1bxoo, buttpark 34\07-74, 5-bromo benzo furan 2-carboxylic acid |
|
C1=CC2=C(C=C1Br)C=C(O2)C(=O)O |
|
241.04 |
|
241.04 |
|
5-Bromo-1-benzofuran-2-carboxylic acid |
Videos
SDS
missing translation for 'documents'