Learn More
5-Bromocytosine, 99%, Thermo Scientific Chemicals
$118.23 - $118.23
Chemical Identifiers
| CAS | 2240-25-7 |
|---|---|
| Molecular Formula | C4H4BrN3O |
| Molecular Weight (g/mol) | 190.00 |
| MDL Number | MFCD00056025 |
| InChI Key | QFVKLKDEXOWFSL-UHFFFAOYSA-N |
| Synonym | 5-bromocytosine, 4-amino-5-bromopyrimidin-2 1h-one, 6-amino-5-bromopyrimidin-2 1h-one, 4-amino-5-bromo-2-pyrimidinol, 4-amino-5-bromopyrimidin-2-ol, 2 1h-pyrimidinone, 4-amino-5-bromo, 4-amino-5-bromo-2-hydroxypyrimidine, 2 1h-pyrimidinone,6-amino-5-bromo, 6-amino-5-bromo-1,2-dihydropyrimidin-2-one, 4-amino-5-bromo-1,2-dihydropyrimidin-2-one |
| PubChem CID | 75233 |
| IUPAC Name | 6-amino-5-bromo-1H-pyrimidin-2-one |
| SMILES | NC1=C(Br)C=NC(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC165782500
|
Thermo Scientific Chemicals
165782500 |
250 mg | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2240-25-7 | |
| 190.00 | |
| QFVKLKDEXOWFSL-UHFFFAOYSA-N | |
| 75233 | |
| NC1=C(Br)C=NC(=O)N1 |
| C4H4BrN3O | |
| MFCD00056025 | |
| 5-bromocytosine, 4-amino-5-bromopyrimidin-2 1h-one, 6-amino-5-bromopyrimidin-2 1h-one, 4-amino-5-bromo-2-pyrimidinol, 4-amino-5-bromopyrimidin-2-ol, 2 1h-pyrimidinone, 4-amino-5-bromo, 4-amino-5-bromo-2-hydroxypyrimidine, 2 1h-pyrimidinone,6-amino-5-bromo, 6-amino-5-bromo-1,2-dihydropyrimidin-2-one, 4-amino-5-bromo-1,2-dihydropyrimidin-2-one | |
| 6-amino-5-bromo-1H-pyrimidin-2-one |
Specifications
| 2240-25-7 | |
| 100.0 | |
| Yellow | |
| 99% | |
| C4H4BrN3O | |
| 250 mg | |
| 5-bromocytosine, 4-amino-5-bromopyrimidin-2 1h-one, 6-amino-5-bromopyrimidin-2 1h-one, 4-amino-5-bromo-2-pyrimidinol, 4-amino-5-bromopyrimidin-2-ol, 2 1h-pyrimidinone, 4-amino-5-bromo, 4-amino-5-bromo-2-hydroxypyrimidine, 2 1h-pyrimidinone,6-amino-5-bromo, 6-amino-5-bromo-1,2-dihydropyrimidin-2-one, 4-amino-5-bromo-1,2-dihydropyrimidin-2-one | |
| NC1=C(Br)C=NC(=O)N1 | |
| 190.00 | |
| 190 | |
| Amorphous Powder |
| 98.5 | |
| 240.0°C to 243.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00056025 | |
| 24, 319 | |
| QFVKLKDEXOWFSL-UHFFFAOYSA-N | |
| 6-amino-5-bromo-1H-pyrimidin-2-one | |
| 75233 | |
| 99% | |
| 5-Bromocytosine, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
If skin irritation occurs: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 218-806-8
RUO – Research Use Only