missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromoindole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107060050

 View more versions of this product

Catalog No. AC107060050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromoindole
10075-50-0
100.0
White to Brown
98.5% min. (GC)
C8H6BrN
5 g
VXWVFZFZYXOBTA-UHFFFAOYSA-N
5-bromo-1H-indole
24905
99%
99%
98.5
89.0°C to 92.0°C
Authentic
Glass bottle
MFCD00005670
5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661
BrC1=CC=C2NC=CC2=C1
196.05
196.05
Chunks and/or Powder

Chemical Identifiers

10075-50-0
196.05
VXWVFZFZYXOBTA-UHFFFAOYSA-N
24905
BrC1=CC=C2NC=CC2=C1
C8H6BrN
MFCD00005670
5-bromoindole, 1h-indole, 5-bromo, 5-bromo indole, 5-bi, 5bromoindole, 5-bromo-indole, zlchem 227, 5-bromo 1h indole, pubchem1661
5-bromo-1H-indole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-208-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only