missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromoisophthalic Acid 98.0+%, TCI America™
$76.43 - $305.70
Chemical Identifiers
| CAS | 23351-91-9 |
|---|---|
| Molecular Formula | C8H3BrO4 |
| Molecular Weight (g/mol) | 243.01 |
| MDL Number | MFCD00152019 |
| InChI Key | JATKASGNRMGFSW-UHFFFAOYSA-L |
| Synonym | 5-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-bromo, 5-bromoisophthalicacid, pubchem22068, 5-bromo-isophthalic acid, acmc-209g2n, ksc202c0n, 5-bromo-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylicacid, 5-bromo |
| PubChem CID | 90073 |
| IUPAC Name | 5-bromobenzene-1,3-dicarboxylate |
| SMILES | [O-]C(=O)C1=CC(=CC(Br)=C1)C([O-])=O |
Chemical Identifiers
| 23351-91-9 | |
| 243.01 | |
| JATKASGNRMGFSW-UHFFFAOYSA-L | |
| 90073 | |
| [O-]C(=O)C1=CC(=CC(Br)=C1)C([O-])=O |
| C8H3BrO4 | |
| MFCD00152019 | |
| 5-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-bromo, 5-bromoisophthalicacid, pubchem22068, 5-bromo-isophthalic acid, acmc-209g2n, ksc202c0n, 5-bromo-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylicacid, 5-bromo | |
| 5-bromobenzene-1,3-dicarboxylate |
Specifications
| 23351-91-9 | |
| White | |
| MFCD00152019 | |
| 5-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-bromo, 5-bromoisophthalicacid, pubchem22068, 5-bromo-isophthalic acid, acmc-209g2n, ksc202c0n, 5-bromo-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylicacid, 5-bromo | |
| [O-]C(=O)C1=CC(=CC(Br)=C1)C([O-])=O | |
| 243.01 | |
| 245.03 | |
| Crystalline Powder |
| 287°C | |
| C8H3BrO4 | |
| 1 g | |
| JATKASGNRMGFSW-UHFFFAOYSA-L | |
| 5-bromobenzene-1,3-dicarboxylate | |
| 90073 | |
| ≥98.0% (GC,T) | |
| 5-Bromoisophthalic Acid |
Safety and Handling
EINECSNumber : (3)-0062
TSCA : Yes