missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-(Bromomethyl)-2,1,3-benzoxadiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC09708DA

 View more versions of this product

Catalog No. CC09708DA


Explore available promotions

Description

Description

Specifications

Specifications

5-(Bromomethyl)-2, 1, 3-benzoxadiazole
96.5
97%
C7H5BrN2O
1 g
BUJFLTNYWUEROF-UHFFFAOYSA-N
5-(bromomethyl)-2,1,3-benzoxadiazole
2735587
97%
32863-31-3
100.0
Amber Glass Bottle
MFCD00174369
5-bromomethyl-2,1,3-benzoxadiazole, 5-bromomethyl benzo c 1,2,5 oxadiazole, 5-bromomethylbenzofurazan, 5-bromomethyl benzofurazan, 2,1,3-benzoxadiazole, 5-bromomethyl, zlchem 1034, 5-bromomethylbenzofurazane, 5-bromomethyl benzo c furazan, 5-bromomethyl-benzo 1,2,5 oxadiazole, 5-bromomethyl-2,1,3-benzooxadiazole
BrCC1=CC2=NON=C2C=C1
213.03
213.03

Chemical Identifiers

32863-31-3
213.03
BUJFLTNYWUEROF-UHFFFAOYSA-N
2735587
BrCC1=CC2=NON=C2C=C1
C7H5BrN2O
MFCD00174369
5-bromomethyl-2,1,3-benzoxadiazole, 5-bromomethyl benzo c 1,2,5 oxadiazole, 5-bromomethylbenzofurazan, 5-bromomethyl benzofurazan, 2,1,3-benzoxadiazole, 5-bromomethyl, zlchem 1034, 5-bromomethylbenzofurazane, 5-bromomethyl benzo c furazan, 5-bromomethyl-benzo 1,2,5 oxadiazole, 5-bromomethyl-2,1,3-benzooxadiazole
5-(bromomethyl)-2,1,3-benzoxadiazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions