Learn More
5-Bromopyridine-2-carboxaldehyde, 99%
CAS: 31181-90-5 | C6H4BrNO | 186.008 g/mol
$116.51 - $430.74
Chemical Identifiers
| CAS | 31181-90-5 |
|---|---|
| Molecular Formula | C6H4BrNO |
| Molecular Weight (g/mol) | 186.008 |
| MDL Number | MFCD04112535 |
| InChI Key | ZQVLPMNLLKGGIU-UHFFFAOYSA-N |
| Synonym | 5-bromopicolinaldehyde, 5-bromo-2-formylpyridine, 5-bromo-2-pyridinecarboxaldehyde, 5-bromopyridine-2-carboxaldehyde, 5-bromo-pyridine-2-carbaldehyde, 5-bromo-2-pyridinecarbaldehyde, 2-pyridinecarboxaldehyde, 5-bromo, 5-bromo-2-pyridine-carbaldehyde, pubchem5059, 5-bromo picolinaldehyde |
| PubChem CID | 9877562 |
| IUPAC Name | 5-bromopyridine-2-carbaldehyde |
| SMILES | C1=CC(=NC=C1Br)C=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5015403
|
Thermo Scientific Chemicals
H5015403 |
1 g |
Each for $116.51
|
|
|||||
|
AAH5015406
|
Thermo Scientific Chemicals
H5015406 |
5 g |
Each for $430.74
|
|
|||||
Description
5-Bromopyridine-2-carboxaldehyde is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromopyridine-2-carboxaldehyde is used as organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
Chemical Identifiers
| 31181-90-5 | |
| 186.008 | |
| ZQVLPMNLLKGGIU-UHFFFAOYSA-N | |
| 9877562 | |
| C1=CC(=NC=C1Br)C=O |
| C6H4BrNO | |
| MFCD04112535 | |
| 5-bromopicolinaldehyde, 5-bromo-2-formylpyridine, 5-bromo-2-pyridinecarboxaldehyde, 5-bromopyridine-2-carboxaldehyde, 5-bromo-pyridine-2-carbaldehyde, 5-bromo-2-pyridinecarbaldehyde, 2-pyridinecarboxaldehyde, 5-bromo, 5-bromo-2-pyridine-carbaldehyde, pubchem5059, 5-bromo picolinaldehyde | |
| 5-bromopyridine-2-carbaldehyde |
Specifications
| 31181-90-5 | |
| C6H4BrNO | |
| 1 g | |
| 5-bromopicolinaldehyde, 5-bromo-2-formylpyridine, 5-bromo-2-pyridinecarboxaldehyde, 5-bromopyridine-2-carboxaldehyde, 5-bromo-pyridine-2-carbaldehyde, 5-bromo-2-pyridinecarbaldehyde, 2-pyridinecarboxaldehyde, 5-bromo, 5-bromo-2-pyridine-carbaldehyde, pubchem5059, 5-bromo picolinaldehyde | |
| ZQVLPMNLLKGGIU-UHFFFAOYSA-N | |
| 5-bromopyridine-2-carbaldehyde | |
| 9877562 | |
| 99% |
| 94°C to 96°C | |
| MFCD04112535 | |
| Air Sensitive | |
| Insoluble in water. | |
| C1=CC(=NC=C1Br)C=O | |
| 186.008 | |
| 186.01 | |
| 5-Bromopyridine-2-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only