Learn More
5-Bromothiophene-2-carboxaldehyde, 97%, Thermo Scientific Chemicals
$63.74 - $577.07
Chemical Identifiers
CAS | 4701-17-1 |
---|---|
Molecular Formula | C5H3BrOS |
Molecular Weight (g/mol) | 191.04 |
MDL Number | MFCD00005432 |
InChI Key | GFBVUFQNHLUCPX-UHFFFAOYSA-N |
Synonym | 5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde |
PubChem CID | 78428 |
IUPAC Name | 5-bromothiophene-2-carbaldehyde |
SMILES | BrC1=CC=C(S1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1338106
|
Thermo Scientific Chemicals
A1338106 |
5 g |
Each for $63.74
|
|
||||||
AAA1338114
|
Thermo Scientific Chemicals
A1338114 |
25 g |
Each for $208.18
|
|
||||||
AAA1338122
|
Thermo Scientific Chemicals
A1338122 |
100 g |
Each for $577.07
|
|
||||||
Description
5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.
Solubility
Soluble in chloroform and methanol.
Notes
Air Sensitive, avoid contact with air/oxygen. Store in cool, dry place in tightly closed container.
Chemical Identifiers
4701-17-1 | |
191.04 | |
GFBVUFQNHLUCPX-UHFFFAOYSA-N | |
78428 | |
BrC1=CC=C(S1)C=O |
C5H3BrOS | |
MFCD00005432 | |
5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde | |
5-bromothiophene-2-carbaldehyde |
Specifications
4701-17-1 | |
104°C to 106°C (10 mmHg) | |
C5H3BrOS | |
MFCD00005432 | |
108404 | |
5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde | |
GFBVUFQNHLUCPX-UHFFFAOYSA-N | |
5-bromothiophene-2-carbaldehyde | |
78428 | |
97% |
1.762 | |
98°C (208°F) | |
1.639 | |
5 g | |
Air sensitive | |
Soluble in chloroform and methanol. | |
BrC1=CC=C(S1)C=O | |
191.04 | |
191.05 | |
5-Bromothiophene-2-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 225-176-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only