missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Bromouracil, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1479922

 View more versions of this product

Catalog No. AAA1479922


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Bromouracil
∼310°C (decomposition)
MFCD00006017
127176
5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
BrC1=CNC(=O)NC1=O
190.98
CHEBI:20552
≥98%
51-20-7
C4H3BrN2O2
100 g
14,1440
LQLQRFGHAALLLE-UHFFFAOYSA-N
5-bromo-1H-pyrimidine-2,4-dione
5802
190.99

Chemical Identifiers

51-20-7
190.98
LQLQRFGHAALLLE-UHFFFAOYSA-N
5802
5-bromo-1H-pyrimidine-2,4-dione
C4H3BrN2O2
MFCD00006017
5-bromouracil, bromouracil, uracil, 5-bromo, 5-bromopyrimidine-2,4 1h,3h-dione, 5-bromo-2,4 1h,3h-pyrimidinedione, 5-bromo-2,4-dihydroxypyrimidine, 2,4 1h,3h-pyrimidinedione, 5-bromo, 5-bromopyrimidine-2,4-diol, unii-4hk400g5uo, 5-bromo-1,2,3,4-tetrahydropyrimidine-2,4-dione
CHEBI:20552
BrC1=CNC(=O)NC1=O
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H341

EINECSNumber : 200-084-0

RTECSNumber : YQ9060000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only