missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2252118

 View more versions of this product

Catalog No. AAB2252118


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone
227°C to 230°C
MFCD02684518
5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one
C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O
251.714
251.71
53786-28-0
C12H14ClN3O
50 g
DOAYWDKFDPSTSV-UHFFFAOYSA-N
6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one
104607
98%

Chemical Identifiers

53786-28-0
251.714
DOAYWDKFDPSTSV-UHFFFAOYSA-N
104607
C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O
C12H14ClN3O
MFCD02684518
5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one
6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 258-771-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only