missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2,4-dinitrobenzoic acid, 98%, Thermo Scientific Chemicals

$230.10 - $1079.27

Chemical Identifiers

CAS 136833-36-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.559
MDL Number MFCD00091829
InChI Key VVQOEJCXWMGEJG-UHFFFAOYSA-N
Synonym 5-chloro-2,4-dinitrobenzoic acid, benzoic acid, 5-chloro-2,4-dinitro, acmc-20a30o, 2,4-dinitro-5-chlorobenzoic acid, 3-chloro-4,6-dinitrobenzoic acid
PubChem CID 3727860
SMILES C1=C(C(=CC(=C1Cl)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2547906
View Documents
Thermo Scientific Chemicals
B2547906
5 g
Each for $230.10
 
AAB2547914
View Documents
Thermo Scientific Chemicals
B2547914
25 g
Each for $1,079.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

136833-36-8
246.559
VVQOEJCXWMGEJG-UHFFFAOYSA-N
3727860
C7H3ClN2O6
MFCD00091829
5-chloro-2,4-dinitrobenzoic acid, benzoic acid, 5-chloro-2,4-dinitro, acmc-20a30o, 2,4-dinitro-5-chlorobenzoic acid, 3-chloro-4,6-dinitrobenzoic acid
C1=C(C(=CC(=C1Cl)[N+](=O)[O-])[N+](=O)[O-])C(=O)O

Specifications

136833-36-8
C7H3ClN2O6
5 g
VVQOEJCXWMGEJG-UHFFFAOYSA-N
246.559
246.56
5-Chloro-2,4-dinitrobenzoic acid
180°C to 182°C
MFCD00091829
5-chloro-2,4-dinitrobenzoic acid, benzoic acid, 5-chloro-2,4-dinitro, acmc-20a30o, 2,4-dinitro-5-chlorobenzoic acid, 3-chloro-4,6-dinitrobenzoic acid
C1=C(C(=CC(=C1Cl)[N+](=O)[O-])[N+](=O)[O-])C(=O)O
3727860
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only