missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloro-2'-deoxyuridine, MP Biomedicals™
$148.38 - $148.38
Chemical Identifiers
CAS | 50-90-8 |
---|---|
Molecular Formula | C9H11ClN2O5 |
Molecular Weight (g/mol) | 262.646 |
InChI Key | NJCXGFKPQSFZIB-XVMARJQXSA-N |
Synonym | 5-chloro-2'-deoxyuridine |
PubChem CID | 13831089 |
IUPAC Name | 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
SMILES | C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10547880
|
MP Biomedicals Inc
0210547880 |
0.1 g |
Each for $148.38
|
|
|||||
Chemical Identifiers
50-90-8 | |
262.646 | |
5-chloro-2'-deoxyuridine | |
5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
C9H11ClN2O5 | |
NJCXGFKPQSFZIB-XVMARJQXSA-N | |
13831089 | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O |
Specifications
50-90-8 | |
>98% | |
0.1 g | |
NJCXGFKPQSFZIB-XVMARJQXSA-N | |
5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
13831089 | |
Powder |
White | |
C9H11ClN2O5 | |
5-chloro-2'-deoxyuridine | |
C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O | |
262.646 | |
262.65 | |
5-Chloro-2'-deoxyuridine |