missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5'-Chloro-2'-hydroxy-3'-nitroacetophenone 98.0+%, TCI America™

Supplier:  TCI America C297325G

 View more versions of this product

Catalog No. C297325G


Explore available promotions

Description

Description

Specifications

Specifications

5′-Chloro-2′-hydroxy-3′-nitroacetophenone
133°C
C8H6ClNO4
25 g
IUNBIQBAYUBIFD-UHFFFAOYSA-N
215.589
215.59
Crystalline Powder
84942-40-5
White-Yellow
MFCD00191921
5'-chloro-2'-hydroxy-3'-nitroacetophenone, 1-5-chloro-2-hydroxy-3-nitrophenyl ethanone, 2-acetyl-6-nitro-4-chlorophenol, 2-acetyl-6-nitro-4-chloro phenol, 2-acetyl-4-chloro-6-nitrophenol, 5-chloro-3-nitro-2-hydroxyacetophenone, benzoic acid,5-chloro-2-hydroxy-3-nitro, ethanone, 1-5-chloro-2-hydroxy-3-nitrophenyl, 1-acetyl-5-chloro-2-hydroxy-3-nitrobenzene, pubchem3355
CC(=O)C1=CC(=CC(=C1O)[N+](=O)[O-])Cl
688237
≥98.0% (GC,T)

Chemical Identifiers

84942-40-5
215.589
IUNBIQBAYUBIFD-UHFFFAOYSA-N
688237
C8H6ClNO4
MFCD00191921
5'-chloro-2'-hydroxy-3'-nitroacetophenone, 1-5-chloro-2-hydroxy-3-nitrophenyl ethanone, 2-acetyl-6-nitro-4-chlorophenol, 2-acetyl-6-nitro-4-chloro phenol, 2-acetyl-4-chloro-6-nitrophenol, 5-chloro-3-nitro-2-hydroxyacetophenone, benzoic acid,5-chloro-2-hydroxy-3-nitro, ethanone, 1-5-chloro-2-hydroxy-3-nitrophenyl, 1-acetyl-5-chloro-2-hydroxy-3-nitrobenzene, pubchem3355
CC(=O)C1=CC(=CC(=C1O)[N+](=O)[O-])Cl
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions