missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Chloro-2-methoxybenzenesulfonyl Chloride 95.0+%, TCI America™
Supplier: TCI America C274025G
Specifications
5-Chloro-2-methoxybenzenesulfonyl Chloride | |
101°C | |
C7H6Cl2O3S | |
25 g | |
5-chloro-2-methoxybenzene-1-sulfonyl chloride, 5-chloro-2-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 5-chloro-2-methoxy, chloro 5-chloro-2-methoxyphenyl sulfone, 5-chloro-2-methoxy-benzenesulfonyl chloride, acmc-1cq3l, ksc494k2b, 4-chloro-2-chlorosulphonyl anisole, 2-methoxy-5-chlorobenzenesulfonyl chloride, 5-chloro-2 methoxybenzenesulfonyl chloride | |
COC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl | |
241.082 | |
241.08 | |
Crystalline Powder |
22952-32-5 | |
White-Yellow | |
MFCD00052959 | |
3261 | |
FCJGLIMDVOTBLO-UHFFFAOYSA-N | |
5-chloro-2-methoxybenzenesulfonyl chloride | |
2734272 | |
≥95.0% (GC,T) |
Chemical Identifiers
22952-32-5 | |
241.082 | |
FCJGLIMDVOTBLO-UHFFFAOYSA-N | |
2734272 | |
COC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl |
C7H6Cl2O3S | |
MFCD00052959 | |
5-chloro-2-methoxybenzene-1-sulfonyl chloride, 5-chloro-2-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 5-chloro-2-methoxy, chloro 5-chloro-2-methoxyphenyl sulfone, 5-chloro-2-methoxy-benzenesulfonyl chloride, acmc-1cq3l, ksc494k2b, 4-chloro-2-chlorosulphonyl anisole, 2-methoxy-5-chlorobenzenesulfonyl chloride, 5-chloro-2 methoxybenzenesulfonyl chloride | |
5-chloro-2-methoxybenzenesulfonyl chloride |
Safety and Handling
TSCA : No