missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-methoxyphenylmagnesium bromide, 0.50M in 2-MeTHF, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H54501AE

Catalog No. AAH54501AE


Explore available promotions

Description

Description

5-Chloro-2-methoxyphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-methoxyphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents
Specifications

Specifications

5-Chloro-2-methoxyphenylmagnesium bromide
C7H6BrClMgO
100 mL
Air and moisture sensitive
It reacts with water.
COC1=CC=C(Cl)C=C1[Mg]Br
245.78
245.79
419535-75-4
MFCD11113495
UN2924
5-chloro-2-methoxyphenylmagnesium bromide, gafvlsuafzzumw-uhfffaoysa-m, bromo 5-chloro-2-methoxyphenyl magnesium, 5-chloro-2-methoxyphenylmagnesium bromide, 0.50m in 2-methf, grignard reagent
GAFVLSUAFZZUMW-UHFFFAOYSA-M
magnesium;1-chloro-4-methoxybenzene-5-ide;bromide
11207325
0.50M in 2-MeTHF

Chemical Identifiers

419535-75-4
245.78
GAFVLSUAFZZUMW-UHFFFAOYSA-M
11207325
COC1=CC=C(Cl)C=C1[Mg]Br
C7H6BrClMgO
MFCD11113495
5-chloro-2-methoxyphenylmagnesium bromide, gafvlsuafzzumw-uhfffaoysa-m, bromo 5-chloro-2-methoxyphenyl magnesium, 5-chloro-2-methoxyphenylmagnesium bromide, 0.50m in 2-methf, grignard reagent
magnesium;1-chloro-4-methoxybenzene-5-ide;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only