missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3291203

 View more versions of this product

Catalog No. AAH3291203


Explore available promotions

Description

Description

5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
Specifications

Specifications

5-Chloro-2-methylbenzenesulfonyl chloride
C7H6Cl2O2S
MFCD09742738
UN3265
Slightly soluble in water.
CC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl
225.083
225.09
34981-38-9
1.5697
1 g
4-chlorotoluene-2-sulphonyl chloride, 4-chlorotoluene-2-sulphonylchloride, 2-chlorosulfonyl-4-chlorotoluene, 5-chloro-2-methylbenzene-1-sulfonyl chloride, 5-chloro-2-methylbenzenesulfonylchloride, benzenesulfonyl chloride, 5-chloro-2-methyl, 5-chloro-2-methylbenzensulfonyl chloride, 5-chloro-2-methylphenylsulfonyl chloride, 3-chloro-6-methyl-benzenesulfonyl chloride, benzenesulfonylchloride, 5-chloro-2-methyl
VHBFBNCERXCECQ-UHFFFAOYSA-N
5-chloro-2-methylbenzenesulfonyl chloride
100415
97%

Chemical Identifiers

34981-38-9
225.083
VHBFBNCERXCECQ-UHFFFAOYSA-N
100415
CC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl
C7H6Cl2O2S
MFCD09742738
4-chlorotoluene-2-sulphonyl chloride, 4-chlorotoluene-2-sulphonylchloride, 2-chlorosulfonyl-4-chlorotoluene, 5-chloro-2-methylbenzene-1-sulfonyl chloride, 5-chloro-2-methylbenzenesulfonylchloride, benzenesulfonyl chloride, 5-chloro-2-methyl, 5-chloro-2-methylbenzensulfonyl chloride, 5-chloro-2-methylphenylsulfonyl chloride, 3-chloro-6-methyl-benzenesulfonyl chloride, benzenesulfonylchloride, 5-chloro-2-methyl
5-chloro-2-methylbenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 252-306-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only