missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-nitroaniline, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 246950500

 View more versions of this product

Catalog No. AC246950500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloro-2-nitroaniline
1635-61-6
100.0
Colorless to Yellow
96% min. (GC)
C6H5ClN2O2
MFCD00007776
12, 730
Solubility in water: 1.22g/L (20°C)
C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]
172.57
172.57
Liquid
97%
96.0
126.0°C to 129.0°C
Authentic
Glass bottle
ClC6H3(NO2)NH2
50 g
2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline
ZCWXYZBQDNFULS-UHFFFAOYSA-N
5-chloro-2-nitroaniline
74218
97%

Chemical Identifiers

1635-61-6
172.57
ZCWXYZBQDNFULS-UHFFFAOYSA-N
74218
C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]
C6H5ClN2O2
MFCD00007776
2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline
5-chloro-2-nitroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-661-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only