Learn More
5-Chloro-2-nitroaniline, 97%, Thermo Scientific Chemicals
$248.81 - $248.81
Chemical Identifiers
CAS | 1635-61-6 |
---|---|
Molecular Formula | C6H5ClN2O2 |
Molecular Weight (g/mol) | 172.57 |
MDL Number | MFCD00007776 |
InChI Key | ZCWXYZBQDNFULS-UHFFFAOYSA-N |
Synonym | 2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline |
PubChem CID | 74218 |
IUPAC Name | 5-chloro-2-nitroaniline |
SMILES | C1=CC(=C(C=C1Cl)N)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC246950500
|
Thermo Scientific Chemicals
246950500 |
50 g | Glass bottle |
Each for $248.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1635-61-6 | |
172.57 | |
ZCWXYZBQDNFULS-UHFFFAOYSA-N | |
74218 | |
C1=CC(=C(C=C1Cl)N)[N+](=O)[O-] |
C6H5ClN2O2 | |
MFCD00007776 | |
2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline | |
5-chloro-2-nitroaniline |
Specifications
1635-61-6 | |
100.0 | |
Colorless to Yellow | |
96% min. (GC) | |
C6H5ClN2O2 | |
MFCD00007776 | |
12, 730 | |
Solubility in water: 1.22g/L (20°C) | |
C1=CC(=C(C=C1Cl)N)[N+](=O)[O-] | |
172.57 | |
172.57 | |
Liquid |
96.0 | |
126.0°C to 129.0°C | |
Authentic | |
Glass bottle | |
ClC6H3(NO2)NH2 | |
50 g | |
2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline | |
ZCWXYZBQDNFULS-UHFFFAOYSA-N | |
5-chloro-2-nitroaniline | |
74218 | |
97% | |
5-Chloro-2-nitroaniline, 97% |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 216-661-5
RUO – Research Use Only