missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

5-Chloro-2-pentanone, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1026922

 View more versions of this product

Catalog No. AAA1026922


Explore available promotions

Description

Description

5-Chloro-2-pentanone used in the preparation of destructible surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-pentanone used in the preparation of destructible surfactants.

Solubility
Soluble in chloroform and methanol.

Notes
keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents
Specifications

Specifications

5-Chloro-2-pentanone
1.053
35°C (95°F)
1.438
100 g
1098490
Soluble in chloroform and methanol.
CC(=O)CCCCl
120.576
120.58
5891-21-4
63°C to 64°C (12 mmHg)
C5H9ClO
MFCD00001008
UN1224
5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one
XVRIEWDDMODMGA-UHFFFAOYSA-N
5-chloropentan-2-one
79993
94%

Chemical Identifiers

5891-21-4
120.576
XVRIEWDDMODMGA-UHFFFAOYSA-N
79993
CC(=O)CCCCl
C5H9ClO
MFCD00001008
5-chloro-2-pentanone, 2-pentanone, 5-chloro, 1-chloro-4-pentanone, 3-chloropropyl methyl ketone, 2-pentanone 5-chloro, 1-chloro-4-oxopentane, pubchem17207, 3-acetylpropyl chloride, 5-chloro-pentan-2-one, 5-chloranylpentan-2-one
5-chloropentan-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 227-565-8

RTECSNumber : SA8143500

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only