missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone ethylene ketal, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 166040250

 View more versions of this product

Catalog No. AC166040250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Chloro-2-pentanone ethylene ketal
5978-08-5
100.0
73°C to 76°C (7.0mmHg)
Authentic
Glass bottle
1.4490 to 1.451
25g
6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate, d-galactose hydrate, dextrose monohydrate, usp, .alpha.-d-glucose hydrate, dextrose, u.s.p.
CC1(CCCCl)OCCO1
164.63
164.63
97%
96.0
1.0900g/mL
72°C
97%
C7H13ClO2
MFCD00003217
1.09
OFERIRWCHSOJJT-UHFFFAOYSA-N
(2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate
133126654
97%

Chemical Identifiers

5978-08-5
164.63
OFERIRWCHSOJJT-UHFFFAOYSA-N
133126654
CC1(CCCCl)OCCO1
C7H13ClO2
MFCD00003217
6-hydroxymethyl oxane-2,3,4,5-tetrol hydrate, d-galactose hydrate, dextrose monohydrate, usp, .alpha.-d-glucose hydrate, dextrose, u.s.p.
(2R,3S,4R,5R,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-776-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only