missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-m-phenylenediamine, 97%

CAS: 33786-89-9 | C6H7ClN2 | 142.586 g/mol

$264.79 - $1099.78

Chemical Identifiers

CAS 33786-89-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
MDL Number MFCD00014775
InChI Key VZNUCJOYPXKLTA-UHFFFAOYSA-N
Synonym 5-chloro-m-phenylenediamine, 3,5-diaminochlorobenzene, 5-chloro-1,3-benzenediamine, 1,3-benzenediamine, 5-chloro, 5-chloro-1,3-phenylenediamine, ccris 2886, 5-amino-3-chloroaniline, acmc-20am42, 2-13-00-00029 beilstein handbook reference, 5-chloro-1,3-diaminobenzene
PubChem CID 73335
IUPAC Name 5-chlorobenzene-1,3-diamine
SMILES C1=C(C=C(C=C1N)Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0648506
View Documents
Thermo Scientific Chemicals
L0648506
5 g
Each for $264.79
Only null left
 
AAL0648514
View Documents
Thermo Scientific Chemicals
L0648514
25 g
Each for $1,099.78
Only null left
 
Description

Description

5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-m-phenylenediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

33786-89-9
142.586
VZNUCJOYPXKLTA-UHFFFAOYSA-N
73335
C1=C(C=C(C=C1N)Cl)N
C6H7ClN2
MFCD00014775
5-chloro-m-phenylenediamine, 3,5-diaminochlorobenzene, 5-chloro-1,3-benzenediamine, 1,3-benzenediamine, 5-chloro, 5-chloro-1,3-phenylenediamine, ccris 2886, 5-amino-3-chloroaniline, acmc-20am42, 2-13-00-00029 beilstein handbook reference, 5-chloro-1,3-diaminobenzene
5-chlorobenzene-1,3-diamine

Specifications

99°C to 103°C
5 g
MFCD00014775
3029854
Slightly soluble in water.
C1=C(C=C(C=C1N)Cl)N
142.586
142.59
5-Chloro-m-phenylenediamine
33786-89-9
C6H7ClN2
UN1673
5-chloro-m-phenylenediamine, 3,5-diaminochlorobenzene, 5-chloro-1,3-benzenediamine, 1,3-benzenediamine, 5-chloro, 5-chloro-1,3-phenylenediamine, ccris 2886, 5-amino-3-chloroaniline, acmc-20am42, 2-13-00-00029 beilstein handbook reference, 5-chloro-1,3-diaminobenzene
VZNUCJOYPXKLTA-UHFFFAOYSA-N
5-chlorobenzene-1,3-diamine
73335
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H341-H302-H312
Toxic if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H341

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES

RTECSNumber : CZ1579500

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only